β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: NADP sodium hydrate, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium monosodium salt xhydrate

A1158555-1G A1158555-5G A1158555-25G
77599-666EA 14.35 USD
77599-666 77599-668 77620-284
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate ≥98%
beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate

Precaución: for Research Use Only

Formula: C₂₁H₂₇N₇NaO₁₇P₃·xH₂O
MDL Number: MFCD00150480
CAS Number: 698999-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR