2,2'-Dimethyl-[1,1'-biphenyl]-4,4'-dicarboxylic acid ≥95%

Supplier: Ambeed
Warning

A1158090-1G A1158090-250MG A1158090-100MG A1158090-5G
77570-962EA 123 USD
77570-962 77595-940 77866-110 77866-111
2,2'-Dimethyl-[1,1'-biphenyl]-4,4'-dicarboxylic acid ≥95%
2,2'-Dimethyl-[1,1'-biphenyl]-4,4'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₄O₄
MDL Number: MFCD31806848
CAS Number: 117490-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR