Benzenesulfonic acid, 4,4',4'',4'''-methanetetrayltetrakis- ≥98%

Supplier: Ambeed
Danger

Synonyms: 4,4',4'',4'''-Methanetetrayltetrabenzenesulphonic acid

A1156852-100MG A1156852-1G
77620-278EA 69.01 USD
77620-278 77620-280
Benzenesulfonic acid, 4,4',4'',4'''-methanetetrayltetrakis- ≥98%
Benzenesulfonic acid, 4,4',4'',4'''-methanetetrayltetrakis-

Precaución: for Research Use Only

Formula: C₂₅H₂₀O₁₂S₄
MW: 640.6791 g/mol
MDL Number: MFCD34165578
CAS Number: 1042222-34-3
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR