Netilmicin sulfate ≥98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4R,5R)-2-(((1S,2S,3R,4S,6R)-4-Amino-3-(((2S,3R)-3-amino-6-(aminomethyl)-3,4-dihydro-2H-pyran-2-yl)oxy)-6-(ethylamino)-2-hydroxycyclohexyl)oxy)-5-methyl-4-(methylamino)tetrahydro-2H-pyran-3,5-diol sulphate(2:5)

A115683-10MG A115683-50MG
77599-662EA 5.13 USD
77599-662 77599-664
Netilmicin sulfate ≥98%
Netilmicin sulfate

Precaución: for Research Use Only

Formula: C₂₁H₄₁N₅O₇·⁵/₂H₂SO₄
MW: 720.75 g/mol
MDL Number: MFCD01311715
CAS Number: 56391-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR