1H-Pyrazole-3-carboxylic acid, 1-(4-carboxyphenyl)- ≥98%

Supplier: Ambeed
Warning

A1156812-1G A1156812-5G
77620-270EA 61.8 USD
77620-270 77866-097
1H-Pyrazole-3-carboxylic acid, 1-(4-carboxyphenyl)- ≥98%
1H-Pyrazole-3-carboxylic acid, 1-(4-carboxyphenyl)-

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₄
MDL Number: MFCD34165566
CAS Number: 72899-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR