2,2'-(2,5-Dimethoxy-1,4-phenylene)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) ≥98%

Supplier: Ambeed
Warning

A1156762-5G A1156762-1G A1156762-250MG
77595-932EA 185.52 USD
77595-932 77620-266 77620-268
2,2'-(2,5-Dimethoxy-1,4-phenylene)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane) ≥98%
2,2'-(2,5-Dimethoxy-1,4-phenylene)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

Precaución: for Research Use Only

Formula: C₂₀H₃₂B₂O₆
MDL Number: MFCD31692685
CAS Number: 1338777-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR