(4-(1,2,2-Triphenylvinyl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1156684-250MG A1156684-5G A1156684-1G
77595-928EA 21.53 USD
77595-928 77595-930 77620-260
(4-(1,2,2-Triphenylvinyl)phenyl)boronic acid ≥98%
(4-(1,2,2-Triphenylvinyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₂₆H₂₁BO₂
MDL Number: MFCD30537204
CAS Number: 1227040-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR