(4,8-Bis(5-(2-ethylhexyl)thiophen-2-yl)benzo[1,2-b:4,5-b']difuran-2,6-diyl)bis(trimethylstannane) ≥99%

Supplier: Ambeed
Danger

A1156411-1G A1156411-250MG
77570-948EA 604 USD
77570-948 77595-924
(4,8-Bis(5-(2-ethylhexyl)thiophen-2-yl)benzo[1,2-b:4,5-b']difuran-2,6-diyl)bis(trimethylstannane) ≥99%
(4,8-Bis(5-(2-ethylhexyl)thiophen-2-yl)benzo[1,2-b:4,5-b']difuran-2,6-diyl)bis(trimethylstannane)

Precaución: for Research Use Only

Formula: C₄₀H₅₈O₂S₂Sn₂
CAS Number: 1391764-83-2
UN: 3146
ADR: 6.1,Ⅰ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR