5-Bromo-4-(2-ethylhexyl)thiophene-2-carbaldehyde ≥99%

Supplier: Ambeed
Warning

A1156328-1G A1156328-100MG A1156328-250MG
77620-250EA 147.29 USD
77620-250 77866-073 77866-074
5-Bromo-4-(2-ethylhexyl)thiophene-2-carbaldehyde ≥99%
5-Bromo-4-(2-ethylhexyl)thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₉BrOS
MDL Number: MFCD31631836
CAS Number: 1356861-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR