3,6-Diethylphthalonitrile ≥99%

Supplier: Ambeed
Danger

A1156325-1G A1156325-100MG A1156325-250MG
77620-248EA 65.92 USD
77620-248 77866-071 77866-072
3,6-Diethylphthalonitrile ≥99%
3,6-Diethylphthalonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
CAS Number: 125773-94-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR