11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide ≥98%

Supplier: Ambeed
Warning

A1155467-1G A1155467-250MG
77783-322EA 184 USD
77783-322 77783-323
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide ≥98%
11-Chloro-1,1'-di-n-propyl-3,3,3',3'-tetramethyl-10,12-trimethyleneindatricarbocyanine iodide

Precaución: for Research Use Only

Formula: C₃₆H₄₄ClIN₂
MDL Number: MFCD00191907
CAS Number: 207399-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR