Cobalt disulfide 99.5% (metals basis excluding Ni) Ni <0.2%
Supplier: Ambeed
|
Danger
|
A1155432-5G
A1155432-25G
A1155432-1G
A1155432-100G
77592-268EA
31.78
USD
77592-268
77595-920
77620-238
77866-065
Cobalt disulfide 99.5% (metals basis excluding Ni) Ni <0.2%
Cobalt disulphide
Precaución: for Research Use Only
Formula:
CoS₂ |
MDL Number:
MFCD00064735 CAS Number: 12013-10-4 UN: 3077 ADR: 9,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...