2,3-Bis(4-bromophenyl)fumaronitrile ≥97%

Supplier: Ambeed
Danger

A1155426-250MG A1155426-1G
77620-236EA 39.14 USD
77620-236 77620-234
2,3-Bis(4-bromophenyl)fumaronitrile ≥97%
2,3-Bis(4-bromophenyl)fumaronitrile

Precaución: for Research Use Only

Formula: C₁₆H₈Br₂N₂
MW: 388.06 g/mol
CAS Number: 82193-93-9
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR