Rhenium oxide 99.9%

Supplier: Ambeed
Danger

A1155331-100MG A1155331-250MG A1155331-5G
77620-228EA 36.05 USD
77620-228 77620-230 77866-064
Rhenium oxide 99.9%
Rhenium oxide

Precaución: for Research Use Only

Formula: O₇Re₂
MDL Number: MFCD00011199
CAS Number: 12624-27-0
UN: 1479
ADR: 5.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR