N-(2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-3-yl)acetamide ≥98%

Supplier: Ambeed
Warning

A1155311-100MG A1155311-1G
77620-226EA 41.2 USD
77620-226 77866-063
N-(2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-3-yl)acetamide ≥98%
N-(2-Oxo-5-(trifluoromethyl)-1,2-dihydropyridin-3-yl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O₂
CAS Number: 1961206-09-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR