MorDalphos Pd G3 ≥98%

Supplier: Ambeed
Warning

A1155299-100MG A1155299-250MG
77620-220EA 35.02 USD
77620-220 77620-222
MorDalphos Pd G3 ≥98%
MorDalphos Pd G3

Precaución: for Research Use Only

Formula: C₄₃H₅₅N₂O₄PPdS
MW: 833.3666 g/mol
MDL Number: MFCD28137452
CAS Number: 2222690-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR