(11bR)-4-Mercaptodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
Supplier: Ambeed
|
Danger
|
A1154900-250MG
A1154900-50MG
77620-200EA
533.54
USD
77620-200
77620-202
(11bR)-4-Mercaptodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥97%
(11bR)-4-Mercaptodinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
Precaución: for Research Use Only
Formula:
C₂₀H₁₃O₃PS MW: 364.3542 g/mol |
CAS Number:
1445903-27-4 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...