1-Benzyl-1-(1-butylpiperidin-4-yl)-3-(3,4-dichlorophenyl)urea hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: NAcM-OPT HCl(2089293-61-6 free base)

A1154860-10MG A1154860-1MG
77620-196EA 50.47 USD
77620-196 77620-198
1-Benzyl-1-(1-butylpiperidin-4-yl)-3-(3,4-dichlorophenyl)urea hydrochloride ≥97%
1-Benzyl-1-(1-butylpiperidin-4-yl)-3-(3,4-dichlorophenyl)urea hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₃₀Cl₃N₃O
MW: 470.86 g/mol
MDL Number: MFCD32202018
CAS Number: 2416146-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR