Cefotiam dihydrochloride ≥98%

Supplier: Ambeed
Danger

Synonyms: (6R,7R)-7-(2-(2-Aminothiazol-4-yl)acetamido)-3-(((1-(2-(dimethylamino)ethyl)-1H-tetrazol-5-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid dihydrochloride, Cefotiam HCl

A115467-1MG A115467-250MG A115467-5G A115467-100MG A115467-10MG A115467-1G A115467-50MG
77685-023EA 5 USD
77685-023 77685-024 77685-025 77901-242 77901-316 77901-377 77901-360
Cefotiam dihydrochloride ≥98%
Cefotiam dihydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₄ClN₉O₄S₃
MDL Number: MFCD09878605
CAS Number: 66309-69-1
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR