Docosahexaenoic acid ethyl ester ≥98%

Supplier: Ambeed
Warning

Synonyms: DHA ethyl ester, Cervonic acid ethyl ester, Ethyl docosahexaenoate, (4Z,7Z,10Z,13Z,16Z,19Z)-Ethyl docosa-4,7,10,13,16,19-hexaenoate

A1154573-100MG A1154573-250MG A1154573-1G A1154573-5G
77599-656EA 21.53 USD
77599-656 77599-658 77595-906 77620-174
Docosahexaenoic acid ethyl ester ≥98%
Docosahexaenoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₂₄H₃₆O₂
MW: 356.54 g/mol
MDL Number: MFCD00673477
CAS Number: 81926-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR