Diethyl (2-cyanoethyl)phosphonate ≥97%

Supplier: Ambeed
Warning

A1154547-1G A1154547-25G A1154547-5G
77866-034EA 52 USD
77866-034 77866-035 77866-036
Diethyl (2-cyanoethyl)phosphonate ≥97%
Diethyl (2-cyanoethyl)phosphonate

Precaución: for Research Use Only

Formula: C₇H₁₄NO₃P
MDL Number: MFCD00013825
CAS Number: 10123-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR