Bis(4-bromophenyl)iodonium trifluoromethanesulfonate ≥97%

Supplier: Ambeed
Danger

Synonyms: Bis(4-bromophenyl)iodonium triflate

A1154333-1G A1154333-25G
77570-912EA 38 USD
77570-912 77866-033
Bis(4-bromophenyl)iodonium trifluoromethanesulfonate ≥97%
Bis(4-bromophenyl)iodonium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₃H₈Br₂F₃IO₃S
MW: 587.97 g/mol
MDL Number: MFCD21608478
CAS Number: 139139-81-4
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR