2-(4-Fluoro-2-nitrophenyl)acetic acid ≥95%

Supplier: Ambeed
Warning

A115344-1G A115344-250MG
77871-477EA 40 USD
77871-477 77871-478
2-(4-Fluoro-2-nitrophenyl)acetic acid ≥95%
2-(4-Fluoro-2-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MDL Number: MFCD04108376
CAS Number: 39616-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR