(2S)-2-Amino-3-((((R)-2,3-bis(palmitoyloxy)propoxy)(hydroxy)phosphoryl)oxy)propanoic acid monosodium salt ≥99%

Supplier: Ambeed
Warning

Synonyms: 12-Dipalmitoyl-sn-glycero-3-PS sodium salt

A1152850-10MG A1152850-1MG A1152850-5MG
77595-894EA 44.08 USD
77595-894 77595-896 77595-898
(2S)-2-Amino-3-((((R)-2,3-bis(palmitoyloxy)propoxy)(hydroxy)phosphoryl)oxy)propanoic acid monosodium salt ≥99%
(2S)-2-Amino-3-((((R)-2,3-bis(palmitoyloxy)propoxy)(hydroxy)phosphoryl)oxy)propanoic acid monosodium salt

Precaución: for Research Use Only

Formula: C₃₈H₇₃NNaO₁₀P
MW: 757.95 g/mol
CAS Number: 145849-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR