(R)-N-(5-(3-Hydroxypyrrolidin-1-yl)-2-morpholinooxazolo[4,5-b]pyridin-6-yl)-2-(2-methylpyridin-4-yl)oxazole-4-carboxamide ≥97%

Supplier: Ambeed
Warning

Synonyms: Emavusertib

A1152397-100MG A1152397-25MG A1152397-50MG
77620-138EA 517.06 USD
77620-138 77620-140 77620-142
(R)-N-(5-(3-Hydroxypyrrolidin-1-yl)-2-morpholinooxazolo[4,5-b]pyridin-6-yl)-2-(2-methylpyridin-4-yl)oxazole-4-carboxamide ≥97%
(R)-N-(5-(3-Hydroxypyrrolidin-1-yl)-2-morpholinooxazolo[4,5-b]pyridin-6-yl)-2-(2-methylpyridin-4-yl)oxazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₄H₂₅N₇O₅
MW: 491.5 g/mol
MDL Number: MFCD32693907
CAS Number: 1801344-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR