2,5-Bis(trifluoromethyl)terephthalic acid ≥97%

Supplier: Ambeed
Warning

A1151681-5G A1151681-25G A1151681-1G
77592-242EA 133.25 USD
77592-242 77620-132 77865-992
2,5-Bis(trifluoromethyl)terephthalic acid ≥97%
2,5-Bis(trifluoromethyl)terephthalic acid

Precaución: for Research Use Only

Formula: C₁₀H₄F₆O₄
MDL Number: MFCD32206254
CAS Number: 366008-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR