N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-methyl-S-(triphenylmethyl)-L-cysteine ≥97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-α-Me-Cys(Trt)-OH

A1151560-100MG A1151560-1G A1151560-250MG
77599-650EA 34.85 USD
77599-650 77599-652 77599-654
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-methyl-S-(triphenylmethyl)-L-cysteine ≥97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-methyl-S-(triphenylmethyl)-L-cysteine

Precaución: for Research Use Only

Formula: C₃₈H₃₃NO₄S
MW: 599.7379 g/mol
MDL Number: MFCD30749166
CAS Number: 725728-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR