(2,2'-Bipyridyl) (2,2'-bis (4-tert-butylpyridine)) ruthenium dichloride ≥98%

Supplier: Ambeed
Warning

A1150987-100MG A1150987-250MG A1150987-1G A1150987-50MG
77620-126EA 256.47 USD
77620-126 77620-128 77865-978 77865-979
(2,2'-Bipyridyl) (2,2'-bis (4-tert-butylpyridine)) ruthenium dichloride ≥98%
(2,2'-Bipyridyl) (2,2'-bis (4-tert-butylpyridine)) ruthenium dichloride

Precaución: for Research Use Only

Formula: C₃₄H₃₆Cl₂N₁₀Ru
MW: 756.69 g/mol
CAS Number: 1821168-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR