Bis (2,2'-bipyridyl) (2,2'-bipyrimidine) ruthenium tetrafluoroborate ≥98%

Supplier: Ambeed
Danger

A1150984-100MG A1150984-250MG A1150984-50MG A1150984-1G
77620-118EA 328.57 USD
77620-118 77620-120 77620-122 77865-975
Bis (2,2'-bipyridyl) (2,2'-bipyrimidine) ruthenium tetrafluoroborate ≥98%
Bis (2,2'-bipyridyl) (2,2'-bipyrimidine) ruthenium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₈H₂₂B₂F₈N₈Ru
CAS Number: 1254690-00-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR