1-(4-sulfobutyl)pyridin-1-ium trifluoromethanesulfonate ≥97%

Supplier: Ambeed
Danger

A1150472-5G A1150472-25G
77865-970EA 48 USD
77865-970 77865-969
1-(4-sulfobutyl)pyridin-1-ium trifluoromethanesulfonate ≥97%
1-(4-sulfobutyl)pyridin-1-ium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₄F₃NO₆S₂
MW: 365.3465 g/mol
MDL Number: MFCD28899127
CAS Number: 855785-75-0
UN: 3265
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR