2-((3,5-Difluoro-3'-methoxy-[1,1'-biphenyl]-4-yl)amino)nicotinic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Farudodstat

A1150468-10MG A1150468-5MG
77595-878EA 58.43 USD
77595-878 77595-880
2-((3,5-Difluoro-3'-methoxy-[1,1'-biphenyl]-4-yl)amino)nicotinic acid ≥97%
2-((3,5-Difluoro-3'-methoxy-[1,1'-biphenyl]-4-yl)amino)nicotinic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₄F₂N₂O₃
MW: 356.32 g/mol
MDL Number: MFCD32215342
CAS Number: 1035688-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR