2-Ethylhexyl propionate ≥97%

Supplier: Ambeed
Warning

A1150320-250MG A1150320-1G A1150320-5G
77570-876EA 36 USD
77570-876 77865-961 77865-962
2-Ethylhexyl propionate ≥97%
2-Ethylhexyl propionate

Precaución: for Research Use Only

Formula: C₁₁H₂₂O₂
MDL Number: MFCD01940631
CAS Number: 6293-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR