Poly(DL-lactide-co-glycolide), M.W. 24000-38000

Supplier: Ambeed
Warning

A1150282-1G A1150282-5G A1150282-250MG A1150282-100MG
77595-874EA 93.28 USD
77595-874 77595-876 77620-108 77865-958
Poly(DL-lactide-co-glycolide), M.W. 24000-38000
Poly(DL-lactide-co-glycolide)

Precaución: for Research Use Only

Formula: [C₃H₄O₂]x[C₂H₂O₂]y
MDL Number: MFCD00131930
CAS Number: 26780-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR