Dibutyltin bis(2-ethylhexanoate) ≥97%

Supplier: Ambeed
Danger

Synonyms: Di-n-butyltin bis(2-ethylhexanoate), Dibutylstannanediyl bis(2-ethylhexanoate)

A1150237-5G A1150237-25G
77620-104EA 6.18 USD
77620-104 77865-956
Dibutyltin bis(2-ethylhexanoate) ≥97%
Dibutyltin bis(2-ethylhexanoate)

Precaución: for Research Use Only

Formula: C₂₄H₄₈O₄Sn
MDL Number: MFCD00015261
CAS Number: 2781-10-4
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR