5-Fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)-N-(5-(piperazin-1-ylmethyl)pyridin-2-yl)pyrimidin-2-amine ≥97%

Supplier: Ambeed
Warning

Synonyms: Abemaciclib metabolite M2

A1149965-1MG A1149965-5MG A1149965-10MG
77595-870EA 59.45 USD
77595-870 77595-872 77620-102
5-Fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)-N-(5-(piperazin-1-ylmethyl)pyridin-2-yl)pyrimidin-2-amine ≥97%
5-Fluoro-4-(4-fluoro-1-isopropyl-2-methyl-1H-benzo[d]imidazol-6-yl)-N-(5-(piperazin-1-ylmethyl)pyridin-2-yl)pyrimidin-2-amine

Precaución: for Research Use Only

Formula: C₂₅H₂₈F₂N₈
MW: 478.54 g/mol
MDL Number: MFCD32067988
CAS Number: 1231930-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR