5,5-Dimethyl-1,3,2-dioxaphosphinane 2-oxide ≥97%

Supplier: Ambeed
Danger

A1149901-1G A1149901-250MG
77595-866EA 126.08 USD
77595-866 77595-868
5,5-Dimethyl-1,3,2-dioxaphosphinane 2-oxide ≥97%
5,5-Dimethyl-1,3,2-dioxaphosphinane 2-oxide

Precaución: for Research Use Only

Formula: C₅H₁₁O₃P
MW: 150.11 g/mol
MDL Number: MFCD03093989
CAS Number: 4090-60-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR