Dihydro cuminyl alcohol ≥89%

Supplier: Ambeed
Warning

Synonyms: Perillyl alcohol, (4-(Prop-1-en-2-yl)cyclohex-1-en-1-yl)methanol

A114980-5G A114980-25G
77570-872EA 16 USD
77570-872 77878-865
Dihydro cuminyl alcohol ≥89%
Dihydro cuminyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₆O
MDL Number: MFCD00001567
CAS Number: 536-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR