1-Methyl-3-(4-sulfobutyl)-1H-imidazol-3-ium trifluoromethanesulfonate ≥97%

Supplier: Ambeed
Danger

A1149492-25G A1149492-5G
77570-866EA 63.86 USD
77570-866 77592-236
1-Methyl-3-(4-sulfobutyl)-1H-imidazol-3-ium trifluoromethanesulfonate ≥97%
1-Methyl-3-(4-sulfobutyl)-1H-imidazol-3-ium trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₉H₁₅F₃N₂O₆S₂
MW: 368.3504 g/mol
CAS Number: 657414-80-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR