Pyrene-1,6-dicarbaldehyde ≥95%

Supplier: Ambeed
Warning

A1149227-100MG A1149227-250MG
77620-072EA 198.79 USD
77620-072 77783-311
Pyrene-1,6-dicarbaldehyde ≥95%
Pyrene-1,6-dicarbaldehyde

Precaución: for Research Use Only

Formula: C₁₈H₁₀O₂
MW: 258.27 g/mol
MDL Number: MFCD32704794
CAS Number: 252338-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR