AMG-510 ≥99%

Supplier: Ambeed
Warning

A1148961-250MG A1148961-100MG
77620-064EA 271.92 USD
77620-064 77865-923
AMG-510 ≥99%
AMG-510

Precaución: for Research Use Only

Formula: C₃₀H₃₀F₂N₆O₃
CAS Number: 2252403-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR