(2S,4R)-1-((S)-2-(3-Aminopropanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC-C2-NH2 2HCl

A1148757-250MG A1148757-100MG
77620-056EA 493.37 USD
77620-056 77620-054
(2S,4R)-1-((S)-2-(3-Aminopropanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride ≥95%
(2S,4R)-1-((S)-2-(3-Aminopropanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₃₇Cl₂N₅O₄S
MW: 574.56 g/mol
CAS Number: 2341796-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR