1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12,15-pentaoxaoctadecan-18-oic acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pomalidomide 4'-PEG5-acid
A1148616-100MG
A1148616-250MG
A1148616-50MG
77595-852EA
291.1
USD
77595-852
77595-854
77595-856
1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12,15-pentaoxaoctadecan-18-oic acid ≥95%
1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12,15-pentaoxaoctadecan-18-oic acid
Precaución: for Research Use Only
Formula:
C₂₆H₃₅N₃O₁₁ MW: 565.57 g/mol |
MDL Number:
MFCD32173790 CAS Number: 2139348-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...