2-Chloro-6-methylpyrimidin-4yl-amine ≥98%

Supplier: Ambeed
Warning

A114858-1G A114858-5G
77747-839EA 16 USD
77747-839 77747-840
2-Chloro-6-methylpyrimidin-4yl-amine ≥98%
2-Chloro-6-methylpyrimidin-4yl-amine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MDL Number: MFCD05863311
CAS Number: 14394-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR