MF-438 95+%

Supplier: Ambeed
Warning

Synonyms: 2-methyl-5-(6-(4-(2-(trifluoromethyl)phenoxy)piperidin-1-yl)pyridazin-3-yl)-1,3,4-thiadiazole

A1148548-10MG A1148548-1MG A1148548-5MG
77595-846EA 223.45 USD
77595-846 77595-848 77595-850
MF-438 95+%
MF-438

Precaución: for Research Use Only

Formula: C₁₉H₁₈F₃N₅OS
MW: 421.44 g/mol
MDL Number: MFCD28386313
CAS Number: 921605-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR