7-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)heptanoic acid ≥95%

Supplier: Ambeed
Warning

A1148289-50MG A1148289-250MG A1148289-1G
77570-856EA 154 USD
77570-856 77595-844 77620-048
7-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)heptanoic acid ≥95%
7-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)heptanoic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₃N₃O₆
MDL Number: MFCD32063493
CAS Number: 2225940-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR