(2S,4R)-1-((S)-2-(5-Aminopentanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: (S,R,S)-AHPC-C4-NH2 HCl

A1148122-100MG A1148122-50MG
77620-036EA 328.57 USD
77620-036 77620-038
(2S,4R)-1-((S)-2-(5-Aminopentanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride ≥95%
(2S,4R)-1-((S)-2-(5-Aminopentanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₇H₄₀ClN₅O₄S
MW: 566.16 g/mol
MDL Number: MFCD31656716
CAS Number: 2245697-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR