Cytidine-5'-monophospho-N-acetylneuraminic acid sodium Salt ≥92%

Supplier: Ambeed
Warning

Synonyms: CMP-Sialic acid sodium salt

A1148020-10MG A1148020-25MG
77595-838EA 55.35 USD
77595-838 77595-840
Cytidine-5'-monophospho-N-acetylneuraminic acid sodium Salt ≥92%
Cytidine-5'-monophospho-N-acetylneuraminic acid sodium Salt

Precaución: for Research Use Only

Formula: C₂₀H₃₀N₄NaO₁₆P
CAS Number: 1007117-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR