6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)hexanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Pomalidomide 4'-alkylC5-acid

A1148000-100MG A1148000-1G A1148000-250MG
77570-852EA 149 USD
77570-852 77865-915 77865-916
6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)hexanoic acid ≥95%
6-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)hexanoic acid

Precaución: for Research Use Only

Formula: C₁₉H₂₁N₃O₆
MDL Number: MFCD32063492
CAS Number: 2225940-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR