tri-Magnesium dicitrate nonahydrate AR

Supplier: Ambeed
Warning

A1147120-100G A1147120-500G A1147120-1000G
77595-820EA 5.13 USD
77595-820 77595-822 77620-008
tri-Magnesium dicitrate nonahydrate AR
tri-Magnesium dicitrate nonahydrate

Precaución: for Research Use Only

Formula: C₁₂H₁₀Mg₃O₁₄·9H₂O
MDL Number: MFCD00036422
CAS Number: 153531-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR