6-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-6-oxohexanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: VH032-linker 5

A1146496-50MG A1146496-100MG A1146496-250MG
77595-816EA 132.23 USD
77595-816 77620-000 77620-002
6-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-6-oxohexanoic acid ≥95%
6-(((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-6-oxohexanoic acid

Precaución: for Research Use Only

Formula: C₂₈H₃₈N₄O₆S
MW: 558.69 g/mol
MDL Number: MFCD32693310
CAS Number: 2172819-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR